Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylbenzylamine 98.0+%, TCI America™
SDP

Catalog No. D45255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45255G 5 g
D45251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45255G Supplier TCI America Supplier No. D45255G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94-98-4
Molecular Formula C9H14N
Molecular Weight (g/mol) 136.22
MDL Number MFCD00025575
InChI Key GBSUVYGVEQDZPG-UHFFFAOYSA-O
Synonym 2,4-dimethylbenzylamine, 2,4-dimethylphenyl methanamine, benzenemethanamine, 2,4-dimethyl, benzylamine, 2,4-dimethyl, 2,4-dimethylbenzenemethanamine, omega-amino-psi-cumene, 1-2,4-dimethylphenyl methanamine, omega-amino-1,2,4-trimethylbenzene, rarechem al bw 0350, timtec-bb sbb005795
PubChem CID 66761
IUPAC Name (2,4-dimethylphenyl)methanaminium
SMILES CC1=CC=C(C[NH3+])C(C)=C1

Specifications

Color Yellow
Boiling Point 219°C
Quantity 5 g
UN Number 2735
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylbenzylamine
EINECSNumber (4)-0286
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.