accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSiClCl

CAS RN 940-41-0

IUPAC Name: trichloro(2-phenylethyl)silane
Synonyms: trichloro(2-phenylethyl)silane
Molecular Weight (g/mol): 239.6
Molecular Formula: C8H9Cl3Si
InChi Key: FMYXZXAKZWIOHO-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)CCC1=CC=CC=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)