Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenethyltrichlorosilane, 95%, Thermo Scientific™

Catalog No. AC432240010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432240010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC432240010 Supplier Thermo Scientific Chemicals Supplier No. 432240010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 940-41-0
Molecular Formula C8H9Cl3Si
Molecular Weight (g/mol) 239.60
MDL Number MFCD00039290
InChI Key FMYXZXAKZWIOHO-UHFFFAOYSA-N
Synonym phenethyltrichlorosilane, trichloro 2-phenylethyl silane, 2-phenylethyl trichlorosilane, silane, trichlorophenethyl, trichlorophenethylsilane, trichloro phenethyl silane, silane, trichloro 2-phenylethyl, 1-phenyl-2-trichlorosilyl ethane, unii-iar3080y7z, trichloro-2-phenylethylsilane
PubChem CID 70327
IUPAC Name trichloro(2-phenylethyl)silane
SMILES Cl[Si](Cl)(Cl)CCC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.2400g/mL
Boiling Point 93°C to 96°C (3.0 mmHg)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5175 to 1.5195 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.24
Formula Weight 239.61
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Phenethyltrichlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store in a dry place.
Store i
EINECSNumber 213-371-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.