accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94089-02-8

IUPAC Name: 1-(thiophen-2-yl)ethane-1-thiol
Synonyms: 1-(thiophen-2-yl)ethanethiol
Molecular Weight (g/mol): 144.25
Molecular Formula: C6H8S2
InChi Key: NCCCTQRHFVSOMP-UHFFFAOYNA-N
SMILES: CC(S)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results