CAS RN 94089-22-2
IUPAC Name:
2-methyl-3-(propan-2-yloxy)pyrazine
Synonyms:
2-isopropoxy-3-methylpyrazine
Molecular Weight (g/mol):
152.2
Molecular Formula:
C8H12N2O
InChi Key:
FRCMVXLVVCXOHO-UHFFFAOYSA-N
SMILES:
CC(C)OC1=NC=CN=C1C
1
–
1
of
1
results
1
2-Methyl-3-isopropoxypyrazine 97.0+%, TCI America™
CAS: 94089-22-2 Molecular Formula: C8H12N2O Molecular Weight (g/mol): 152.197 MDL Number: MFCD00059772 InChI Key: FRCMVXLVVCXOHO-UHFFFAOYSA-N Synonym: 2-Isopropoxy-3-methylpyrazine PubChem CID: 3086387 IUPAC Name: 2-methyl-3-propan-2-yloxypyrazine SMILES: CC1=NC=CN=C1OC(C)C