CAS RN 942425-68-5
IUPAC Name:
2-(pyridin-4-yl)-1H,4H,5H,6H,7H-pyrrolo[3,2-c]pyridin-4-one hydrochloride
Synonyms:
2-(pyridin-4-yl)-1H,5H,6H,7H-pyrrolo[3,2-c]pyridin-4-one hydrochloride
Molecular Weight (g/mol):
249.7
Molecular Formula:
C12H12ClN3O
InChi Key:
IMVNFURYBZMFDZ-UHFFFAOYSA-N
SMILES:
Cl.O=C1NCCC2=C1C=C(N2)C1=CC=NC=C1
1
–
1
of
1
results
1
R&D Systems™ PHA 767491 hydrochloride
CAS: 942425-68-5 Molecular Formula: C12H12ClN3O