accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94402-42-3

OOBr

CAS RN 94402-42-3

IUPAC Name: 2-bromo-1-(2-phenoxyphenyl)ethan-1-one
Synonyms: 2-bromo-1-(2-phenoxyphenyl)ethanone
Molecular Weight (g/mol): 291.14
Molecular Formula: C14H11BrO2
InChi Key: UWRDQDVDGPRCKR-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=CC=C1OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results