accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNHNOHNN

CAS RN 944328-88-5

IUPAC Name: 2-{[9-({[1,1'-biphenyl]-4-yl}methyl)-2-(2,3-dihydro-1H-inden-5-yloxy)-9H-purin-6-yl]amino}-3-phenylpropan-1-ol
Synonyms: 2-[(9-{[1,1'-biphenyl]-4-ylmethyl}-2-(2,3-dihydro-1H-inden-5-yloxy)purin-6-yl)amino]-3-phenylpropan-1-ol
Molecular Weight (g/mol): 567.69
Molecular Formula: C36H33N5O2
InChi Key: DOKZLKDGUQWMSX-UHFFFAOYNA-N
SMILES: OCC(CC1=CC=CC=C1)NC1=C2N=CN(CC3=CC=C(C=C3)C3=CC=CC=C3)C2=NC(OC2=CC3=C(CCC3)C=C2)=N1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results