CAS RN 944342-90-9
IUPAC Name:
N4-[4-(benzyloxy)-3-chlorophenyl]-5-({[2-(morpholin-4-yl)ethoxy]imino}methyl)pyrimidine-4,6-diamine hydrochloride
Synonyms:
N4-[4-(benzyloxy)-3-chlorophenyl]-5-({[2-(morpholin-4-yl)ethoxy]imino}methyl)pyrimidine-4,6-diamine hydrochloride
Molecular Weight (g/mol):
519.43
Molecular Formula:
C24H28Cl2N6O3
InChi Key:
ZXKZRKQMKNRZNN-UHFFFAOYSA-N
SMILES:
Cl.NC1=C(C=NOCCN2CCOCC2)C(NC2=CC(Cl)=C(OCC3=CC=CC=C3)C=C2)=NC=N1
1
–
1
of
1
results
1
R&D Systems™ JNJ 28871063 hydrochloride
CAS: 944342-90-9 Molecular Formula: C24H28Cl2N6O3