accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CCH3POOOOPCH3CH3CH3H3CH3CCH3CH3ClRuCl(?)

CAS RN 944451-30-3

IUPAC Name: (4-{5-[bis(3,5-dimethylphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-dimethylphenyl)phosphane; 1-methyl-4-(propan-2-yl)benzene; dichlororuthenium
Synonyms: (4-{5-[bis(3,5-dimethylphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-dimethylphenyl)phosphane; cymene; dichlororuthenium
Molecular Weight (g/mol): 1028.99
Molecular Formula: C56H58Cl2O4P2Ru
InChi Key: LLNJPFQWQJQPEH-UHFFFAOYSA-L
SMILES: Cl[Ru]Cl.CC(C)C1=CC=C(C)C=C1.CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=C(C2=C(OCO2)C=C1)C1=C(C=CC2=C1OCO2)P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results