accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 94465-25-5

IUPAC Name: 9-(4-methoxyphenyl)-9H-xanthen-9-ol
Synonyms: 9-(4-methoxyphenyl)xanthen-9-ol
Molecular Weight (g/mol): 304.35
Molecular Formula: C20H16O3
InChi Key: ZYACEJZTEMQQEU-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1(O)C2=CC=CC=C2OC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results