accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOHON(R)CH3

CAS RN 944797-51-7

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-3-[(triphenylmethyl)sulfanyl]propanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}-3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 599.75
Molecular Formula: C38H33NO4S
InChi Key: RAKOPMQMPUNRGI-DHUJRADRSA-N
SMILES: CN([C@@H](CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results