accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONO

CAS RN 945128-26-7

IUPAC Name: N,N-dicyclohexyl-5-cyclopropyl-1,2-oxazole-3-carboxamide
Synonyms: N,N-dicyclohexyl-5-cyclopropyl-1,2-oxazole-3-carboxamide
Molecular Weight (g/mol): 316.45
Molecular Formula: C19H28N2O2
InChi Key: NDKGACIWVAOUQH-UHFFFAOYSA-N
SMILES: O=C(N(C1CCCCC1)C1CCCCC1)C1=NOC(=C1)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)