accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CO(R)(S)HONON

CAS RN 94535-50-9

IUPAC Name: (3S,4R)-3-hydroxy-2,2-dimethyl-4-(2-oxopyrrolidin-1-yl)-3,4-dihydro-2H-1-benzopyran-6-carbonitrile
Synonyms: (-)-cromakalim lemakalim levcromakalim levcromakelim
Molecular Weight (g/mol): 286.33
Molecular Formula: C16H18N2O3
InChi Key: TVZCRIROJQEVOT-CABCVRRESA-N
SMILES: CC1(C)OC2=CC=C(C=C2[C@H]([C@@H]1O)N1CCCC1=O)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results