accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94593-08-5

IUPAC Name: 3,3,5,5-tetraphenyl-2,4,6-trioxa-3,5-disilaheptane
Synonyms: 3,3,5,5-tetraphenyl-2,4,6-trioxa-3,5-disilaheptane
Molecular Weight (g/mol): 442.66
Molecular Formula: C26H26O3Si2
InChi Key: CDHRQJOOOVAAFQ-UHFFFAOYSA-N
SMILES: CO[Si](O[Si](OC)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results