accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NHNCH3H3CSOOClCl

CAS RN 946387-07-1

IUPAC Name: 2,4-dichloro-N-(propan-2-yl)-N-{2-[(propan-2-yl)amino]ethyl}benzene-1-sulfonamide
Synonyms: 2,4-dichloro-N-isopropyl-N-[2-(isopropylamino)ethyl]benzenesulfonamide
Molecular Weight (g/mol): 353.3
Molecular Formula: C14H22Cl2N2O2S
InChi Key: IHYZMEAZAIFMTN-UHFFFAOYSA-N
SMILES: CC(C)NCCN(C(C)C)S(=O)(=O)C1=C(Cl)C=C(Cl)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results