accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3ONOH3CNH

CAS RN 947303-87-9

IUPAC Name: 4-[(diethylamino)methyl]-N-[2-(2-methoxyphenyl)ethyl]-N-(pyrrolidin-3-yl)benzamide
Synonyms: 4-[(diethylamino)methyl]-N-[2-(2-methoxyphenyl)ethyl]-N-(pyrrolidin-3-yl)benzamide
Molecular Weight (g/mol): 409.57
Molecular Formula: C25H35N3O2
InChi Key: XKPJTOHUPQWSOJ-UHFFFAOYNA-N
SMILES: CCN(CC)CC1=CC=C(C=C1)C(=O)N(CCC1=CC=CC=C1OC)C1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)