CAS RN 94787-08-3
IUPAC Name:
1,3-benzothiazol-2-amine hydrochloride
Synonyms:
2-aminobenzothiazole hydrochloride
Molecular Weight (g/mol):
186.66
Molecular Formula:
C7H7ClN2S
InChi Key:
UEQOZTASNGWDPC-UHFFFAOYSA-N
SMILES:
Cl.NC1=NC2=CC=CC=C2S1
1
–
1
of
1
results
1
2-Aminobenzothiazole Hydrochloride 98.0+%, TCI America™
CAS: 94787-08-3 Molecular Formula: C7H7ClN2S Molecular Weight (g/mol): 186.657 MDL Number: MFCD00142859 InChI Key: UEQOZTASNGWDPC-UHFFFAOYSA-N PubChem CID: 3027575 IUPAC Name: 1,3-benzothiazol-2-amine;hydrochloride SMILES: C1=CC=C2C(=C1)N=C(S2)N.Cl