accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3OOOCH3(E/Z)(?)

CAS RN 94856-25-4

IUPAC Name: 3-(adamantan-2-ylidene)-4-[1-(2,5-dimethylfuran-3-yl)ethylidene]oxolane-2,5-dione
Synonyms: 3-(adamantan-2-ylidene)-4-[1-(2,5-dimethylfuran-3-yl)ethylidene]oxolane-2,5-dione
Molecular Weight (g/mol): 352.43
Molecular Formula: C22H24O4
InChi Key: OOVNPWVABCFZRY-UHFFFAOYSA-N
SMILES: CC(C1=C(C)OC(C)=C1)=C1C(=O)OC(=O)C1=C1C2CC3CC(C2)CC1C3
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results