Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aberchrome 670 98.0+%, TCI America™
SDP

Catalog No. A2869200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2869200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2869200MG Supplier TCI America Supplier No. A2869200MG

Specifications

CAS 94856-25-4
Melting Point 181°C
Molecular Formula C22H24O4
Quantity 200 mg
MDL Number MFCD07368579
Synonym (E)-3-(Adamantan-2-ylidene)-4-[1-(2,5-dimethyl-3-furyl)ethylidene]dihydro-2,5-furandione
InChI Key OOVNPWVABCFZRY-UHFFFAOYSA-N
SMILES CC(C1=C(C)OC(C)=C1)=C1C(=O)OC(=O)C1=C1C2CC3CC(C2)CC1C3
IUPAC Name 3-(adamantan-2-ylidene)-4-[1-(2,5-dimethylfuran-3-yl)ethylidene]oxolane-2,5-dione
Molecular Weight (g/mol) 352.43
PubChem CID 15936866
Formula Weight 352.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Aberchrome 670
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.