accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNH2NFHClHCl

CAS RN 950912-80-8

IUPAC Name: 4-[1-cyclohexyl-4-(4-fluorophenyl)-1H-imidazol-5-yl]pyrimidin-2-amine dihydrochloride
Synonyms: 4-[3-cyclohexyl-5-(4-fluorophenyl)imidazol-4-yl]pyrimidin-2-amine dihydrochloride
Molecular Weight (g/mol): 410.32
Molecular Formula: C19H22Cl2FN5
InChi Key: PSNKGVAXBSAHCH-UHFFFAOYSA-N
SMILES: Cl.Cl.NC1=NC=CC(=N1)C1=C(N=CN1C1CCCCC1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results