CAS RN 951322-11-5
IUPAC Name:
{2-[6-bromo-2-(2-methylbut-3-en-2-yl)-1H-indol-3-yl]ethyl}(methyl)amine hydrochloride
Synonyms:
desformylflustrabromine
Molecular Weight (g/mol):
357.72
Molecular Formula:
C16H22BrClN2
InChi Key:
GEZWEAPBLKXKGN-UHFFFAOYSA-N
SMILES:
Cl.CNCCC1=C(NC2=CC(Br)=CC=C12)C(C)(C)C=C
1
–
1
of
1
results
1
R&D Systems™ Desformylflustrabromine hydrochloride
CAS: 951322-11-5 Molecular Formula: C16H22BrClN2