accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CCH3FFFSOONSOOFFF

CAS RN 952155-74-7

IUPAC Name: N,N,N-trimethylcyclohexanaminium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: N,N,N-trimethylcyclohexanaminium; bistriflylimide anion
Molecular Weight (g/mol): 422.4
Molecular Formula: C11H20F6N2O4S2
InChi Key: UFYPPIXBMHRHIA-UHFFFAOYSA-N
SMILES: C[N+](C)(C)C1CCCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results