accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95241-36-4

IUPAC Name: 2-(propan-2-yloxy)ethyl benzoate
Synonyms: 2-isopropoxyethyl benzoate
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: QLSUGJQAAUGJHE-UHFFFAOYSA-N
SMILES: CC(C)OCCOC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results