Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropoxyethyl Benzoate 95.0+%, TCI America™
SDP

Catalog No. B09725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09725ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B09725ML Supplier TCI America Supplier No. B09725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95241-36-4
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00059357
InChI Key QLSUGJQAAUGJHE-UHFFFAOYSA-N
Synonym Benzoic Acid 2-Isopropoxyethyl Ester, Ethylene Glycol Monoisopropyl Ether Benzoate
PubChem CID 570438
IUPAC Name 2-propan-2-yloxyethyl benzoate
SMILES CC(C)OCCOC(=O)C1=CC=CC=C1

Specifications

Color Colorless
Quantity 5 mL
Formula Weight 208.26
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isopropoxyethyl Benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.