accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CN

CAS RN 952431-30-0

IUPAC Name: N-{[1,1'-biphenyl]-4-yl}-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-[1,1'-biphenyl]-4-amine
Synonyms: N-{[1,1'-biphenyl]-4-yl}-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 523.48
Molecular Formula: C36H34BNO2
InChi Key: PESBFNLGEMHNAM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results