accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNNHNNNHSNHFFFFFF

CAS RN 956014-19-0

IUPAC Name: 1-[3,5-bis(trifluoromethyl)phenyl]-3-[4-bromo-2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]thiourea
Synonyms: 1-[3,5-bis(trifluoromethyl)phenyl]-3-[4-bromo-2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]thiourea
Molecular Weight (g/mol): 511.24
Molecular Formula: C16H9BrF6N6S
InChi Key: MDKAFDIKYQMOMF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(NC(=S)NC2=C(C=C(Br)C=C2)C2=NNN=N2)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)