CAS RN 956579-01-4
CAS RN 956579-01-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ IRDye™ 800CW, NHS ester
Near-infared dye (reactive group NHS ester)