Learn More
Description
Application: Fluorescence microscopy.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 773 nm and 792 nm, respectively.
Please see the protocol for further information on protein/antibody labeling and conjugation.
It is recommended to dissolve this dye in water or DMSO (10 - 20 mg/mL)
Specifications
Specifications
| Chemical Name or Material | Sodium 2-((E)-2-((E)-3-(2-((E)-1-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-3,3-dimethyl-5-sulfonatoindolin-2-ylidene)ethylidene)-2-(4-sulfonatophenoxy)cyclohex-1-en-1-yl)vinyl)-3,3-dimethyl-1-(4-sulfonatobutyl)-3H-indol-1-ium-5-sulfonate |
| CAS | 956579-01-4 |
| Quantity | 500 μg |
| Target | Fluorescent Dyes |
| Molecular Formula | C50H54N3Na3O17S4 |
| Purity | 0.8 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
