accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOHONNNNNNNOHN

CAS RN 956906-93-7

IUPAC Name: 2-(4-{1-[(quinolin-6-yl)methyl]-1H-[1,2,3]triazolo[4,5-b]pyrazin-6-yl}-1H-pyrazol-1-yl)ethan-1-ol; methanesulfonic acid
Synonyms: 2-{4-[3-(quinolin-6-ylmethyl)-[1,2,3]triazolo[4,5-b]pyrazin-5-yl]pyrazol-1-yl}ethanol; methanesulfonic acid
Molecular Weight (g/mol): 468.49
Molecular Formula: C20H20N8O4S
InChi Key: HBEMHKVWZJTVOC-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.OCCN1C=C(C=N1)C1=CN=C2N=NN(CC3=CC4=CC=CN=C4C=C3)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)