accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHN

CAS RN 959-66-0

IUPAC Name: 3-oxo-N,3-diphenylpropanamide
Synonyms: 3-oxo-N,3-diphenylpropanamide
Molecular Weight (g/mol): 239.27
Molecular Formula: C15H13NO2
InChi Key: XRZDIHADHZSFBB-UHFFFAOYSA-N
SMILES: O=C(CC(=O)C1=CC=CC=C1)NC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)