Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzoylacetanilide 98%, Thermo Scientific™

Catalog No. AC154080250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC154080250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC154080250 Supplier Thermo Scientific Chemicals Supplier No. 154080250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-66-0
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.27
MDL Number MFCD00003084
InChI Key XRZDIHADHZSFBB-UHFFFAOYSA-N
Synonym 2-benzoylacetanilide, benzoylacetanilide, 3-oxo-3-phenylpropionanilide, benzenepropanamide, .beta.-oxo-n-phenyl, acetamide,n-2-benzoylphenyl, 3-oxo-3-phenyl-n-phenylpropanamide, alpha-benzoylacetanilide, benzoylacetoanilide, .alpha.-benzoylacetanilide, .alpha.-benzoylacetoanilide
PubChem CID 70398
IUPAC Name 3-oxo-N,3-diphenylpropanamide
SMILES O=C(CC(=O)C1=CC=CC=C1)NC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 105°C to 109°C
Color Yellow
Infrared Spectrum Authentic
Quantity 25 g
Beilstein 12, 522
Formula Weight 239.27
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 2-Benzoylacetanilide, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.