Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzoylacetanilide 98.0+%, TCI America™
SDP

Catalog No. B090425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B090425G 25 g
B09045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B090425G Supplier TCI America Supplier No. B090425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 959-66-0
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.27
MDL Number MFCD00003084
InChI Key XRZDIHADHZSFBB-UHFFFAOYSA-N
Synonym 2-benzoylacetanilide, benzoylacetanilide, 3-oxo-3-phenylpropionanilide, benzenepropanamide, .beta.-oxo-n-phenyl, acetamide,n-2-benzoylphenyl, 3-oxo-3-phenyl-n-phenylpropanamide, alpha-benzoylacetanilide, benzoylacetoanilide, .alpha.-benzoylacetanilide, .alpha.-benzoylacetoanilide
PubChem CID 70398
IUPAC Name 3-oxo-N,3-diphenylpropanamide
SMILES O=C(CC(=O)C1=CC=CC=C1)NC1=CC=CC=C1

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 25 g
Formula Weight 239.27
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzoylacetanilide
EINECSNumber (3)-1388
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.