accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95915-82-5

IUPAC Name: N-(6-aminohexyl)-2-nitrobenzene-1-sulfonamide hydrochloride
Synonyms: N-(6-aminohexyl)-2-nitrobenzenesulfonamide hydrochloride
Molecular Weight (g/mol): 337.82
Molecular Formula: C12H20ClN3O4S
InChi Key: CRQMJHQZDIEKGD-UHFFFAOYSA-N
SMILES: Cl.NCCCCCCNS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results