accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClBrBr

CAS RN 96-12-8

IUPAC Name: 1,2-dibromo-3-chloropropane
Synonyms: 1,2-dibromo-3-chloropropane nemagon soil fumigant nemagon 20G nemagon 206 caswell no. 287 nematocide EM 12.1 nemabrom nematocide EM 15.1 fumazon 86 fumagon
Molecular Weight (g/mol): 236.33
Molecular Formula: C3H5Br2Cl
InChi Key: WBEJYOJJBDISQU-UHFFFAOYNA-N
SMILES: ClCC(Br)CBr
Solvent or Matrix 
  • (1)
  • (1)
Element 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)