Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromo-3-chloropropane Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USHC3401
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-HC-340-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-HC-340-1 Supplier Agilent Technologies Supplier No. HC3401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 96-12-8 , 67-56-1
Molecular Formula C3H5Br2Cl
Molecular Weight (g/mol) 236.33
MDL Number MFCD00039365
InChI Key WBEJYOJJBDISQU-UHFFFAOYNA-N
PubChem CID 7280
ChEBI CHEBI:34044
IUPAC Name 1,2-dibromo-3-chloropropane
SMILES ClCC(Br)CBr

Specifications

CAS Max % 99.98, 0.01
Analyte or Component Names Methanol,1,2-dibromo-3-chloropropane (DBCP)
Element Methanol, 1,2-Dibromo-3-chloropropane
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material 1, 2-Dibromo-3-chloropropane Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.