Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromo-3-chloropropane 98.0+%, TCI America™
SDP

Catalog No. C080625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C080625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C080625G Supplier TCI America Supplier No. C080625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96-12-8
Molecular Formula C3H5Br2Cl
Molecular Weight (g/mol) 236.33
MDL Number MFCD00039365
InChI Key WBEJYOJJBDISQU-UHFFFAOYNA-N
Synonym 3-chloro-1,2-dibromopropane, dbcp, fumazone, nemagon, dibromochloropropane, fumagon, nemabrom, nemafume, nemanax, nemapaz
PubChem CID 7280
ChEBI CHEBI:34044
IUPAC Name 1,2-dibromo-3-chloropropane
SMILES ClCC(Br)CBr

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 200°C
Quantity 25 g
UN Number 2872
Formula Weight 236.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dibromo-3-chloropropane
EINECSNumber (2)-0082
RTECSNumber TX8750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.