accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3

CAS RN 96-37-7

IUPAC Name: methylcyclopentane
Synonyms: methylcyclopentane
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: GDOPTJXRTPNYNR-UHFFFAOYSA-N
SMILES: CC1CCCC1
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Boiling Point 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)