accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 961-68-2

ONONHNOO

CAS RN 961-68-2

IUPAC Name: 2,4-dinitro-N-phenylaniline
Synonyms: 2,4-dinitro-N-phenylaniline
Molecular Weight (g/mol): 259.22
Molecular Formula: C12H9N3O4
InChi Key: RHTVQEPJVKUMPI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(NC2=CC=CC=C2)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)