Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrodiphenylamine 98.0+%, TCI America™
SDP

Catalog No. D027825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D027825G 25 g
D0278100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D027825G Supplier TCI America Supplier No. D027825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 961-68-2
Molecular Formula C12H9N3O4
Molecular Weight (g/mol) 259.221
MDL Number MFCD00007223
InChI Key RHTVQEPJVKUMPI-UHFFFAOYSA-N
Synonym 2,4-dinitrodiphenylamine, serisol yellow 2g, supracet yellow 3g, c.i. disperse yellow 14, o,p-dinitrodiphenylamine, acetoquinone yellow 5jz, benzenamine, 2,4-dinitro-n-phenyl, n-phenyl-2,4-dinitroaniline, diphenylamine, 2,4-dinitro, n-2,4-dinitrophenyl aniline
PubChem CID 13748
IUPAC Name 2,4-dinitro-N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 157°C
Quantity 25 g
Formula Weight 259.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitrodiphenylamine
RTECSNumber JJ8825000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.