Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrodiphenylamine, MP Biomedicals

Catalog No. ICN205756 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205756 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205756 Supplier MP Biomedicals, Inc Supplier No. 0520575610

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 961-68-2
Molecular Formula C12H9N3O4
Molecular Weight (g/mol) 259.221
InChI Key RHTVQEPJVKUMPI-UHFFFAOYSA-N
Synonym 2,4-dinitrodiphenylamine, serisol yellow 2g, supracet yellow 3g, c.i. disperse yellow 14, o,p-dinitrodiphenylamine, acetoquinone yellow 5jz, benzenamine, 2,4-dinitro-n-phenyl, n-phenyl-2,4-dinitroaniline, diphenylamine, 2,4-dinitro, n-2,4-dinitrophenyl aniline
PubChem CID 13748
IUPAC Name 2,4-dinitro-N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 10 g
Formula Weight 259.21
Chemical Name or Material 2, 4-Dinitrodiphenylamine
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.