accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHN

CAS RN 96163-72-3

IUPAC Name: ethyl pyrrolidine-2-carboxylate
Synonyms: ethyl pyrrolidine-2-carboxylate
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: QPNJHVDIRZNKOX-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCN1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results