Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Proline Ethyl Ester, TCI America™
SDP

Catalog No. P23265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23265G 5 g
P23261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23265G Supplier TCI America Supplier No. P23265G

Chemical Identifiers

CAS 96163-72-3
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD17271681
InChI Key QPNJHVDIRZNKOX-ZCFIWIBFSA-N
Synonym H-D-Pro-OEt
PubChem CID 11499235
IUPAC Name ethyl (2R)-pyrrolidine-2-carboxylate
SMILES CCOC(=O)C1CCCN1

Specifications

Color Yellow
Boiling Point 74°C
Quantity 5 g
Formula Weight 143.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material D-Proline Ethyl Ester
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.