accessibility menu, dialog, popup

UserName

Viewing profile as user:
IOOH2O

CAS RN 96195-89-0

IUPAC Name: (2-carboxylatophenyl)(phenyl)iodanium hydrate
Synonyms: (2-carboxylatophenyl)(phenyl)iodanium hydrate
Molecular Weight (g/mol): 342.13
Molecular Formula: C13H11IO3
InChi Key: NHCFFVMWVHFFSJ-UHFFFAOYSA-N
SMILES: O.[O-]C(=O)C1=CC=CC=C1[I+]C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results