CAS RN 965-52-6
IUPAC Name:
4-hydroxy-N'-[(E)-(5-nitrofuran-2-yl)methylidene]benzohydrazide
Synonyms:
akabar
Molecular Weight (g/mol):
275.22
Molecular Formula:
C12H9N3O5
InChi Key:
YCWSUKQGVSGXJO-NTUHNPAUSA-N
SMILES:
OC1=CC=C(C=C1)C(=O)N\N=C\C1=CC=C(O1)[N+]([O-])=O
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Nifuroxazide
A cell-permeable and orally available nitrofuran-based antidiarrheal agent that effectively suppresses the activation of cellular STAT1/3/5 transcription activity.