CAS RN 96530-81-3
IUPAC Name:
4-fluoro-2-methoxypyridine
Synonyms:
4-fluoro-2-methoxypyridine
Molecular Weight (g/mol):
127.12
Molecular Formula:
C6H6FNO
InChi Key:
RGWFJFBUHTUVMV-UHFFFAOYSA-N
SMILES:
COC1=NC=CC(F)=C1
1
–
1
of
1
results
1
4-Fluoro-2-methoxypyridine, 97%, Thermo Scientific™
CAS: 96530-81-3 Molecular Formula: C6H6FNO Molecular Weight (g/mol): 127.12 MDL Number: MFCD09835267 InChI Key: RGWFJFBUHTUVMV-UHFFFAOYSA-N Synonym: pyridine, 4-fluoro-2-methoxy,4-fluoro-2-methoxy-pyridine,4-fluoro 2-methoxy pyridine,2-methoxy-4-fluoropyridine,fluoromethoxypyridine,pubchem6596,acmc-209v4d,4-fluoranyl-2-methoxy-pyridine,3,3'-methylenebis 5-methyloxazolidine,pyridine, 4-fluoro-2-methoxy-9ci PubChem CID: 338458 IUPAC Name: 4-fluoro-2-methoxypyridine SMILES: COC1=NC=CC(F)=C1