Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-methoxypyridine, 97%, Thermo Scientific™

Catalog No. AC450010010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450010010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC450010010 Supplier Thermo Scientific Chemicals Supplier No. 450010010
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 96530-81-3
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.12
MDL Number MFCD09835267
InChI Key RGWFJFBUHTUVMV-UHFFFAOYSA-N
Synonym pyridine, 4-fluoro-2-methoxy, 4-fluoro-2-methoxy-pyridine, 4-fluoro 2-methoxy pyridine, 2-methoxy-4-fluoropyridine, fluoromethoxypyridine, pubchem6596, acmc-209v4d, 4-fluoranyl-2-methoxy-pyridine, 3,3'-methylenebis 5-methyloxazolidine, pyridine, 4-fluoro-2-methoxy-9ci
PubChem CID 338458
IUPAC Name 4-fluoro-2-methoxypyridine
SMILES COC1=NC=CC(F)=C1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 127.12
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methoxypyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.