accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 97-07-4

IUPAC Name: 1-chloro-4-methanesulfonyl-2-nitrobenzene
Synonyms: 1-chloro-4-methanesulfonyl-2-nitrobenzene
Molecular Weight (g/mol): 235.64
Molecular Formula: C7H6ClNO4S
InChi Key: JAANTSGNTKWLFA-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results