Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrophenyl Methyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. C27595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27595G Supplier TCI America Supplier No. C27595G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97-07-4
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.638
MDL Number MFCD00039754
InChI Key JAANTSGNTKWLFA-UHFFFAOYSA-N
PubChem CID 66796
IUPAC Name 1-chloro-4-methylsulfonyl-2-nitrobenzene
SMILES CS(=O)(=O)C1=CC(=C(C=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 125°C
Color Yellow
Quantity 5 g
Formula Weight 235.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrophenyl Methyl Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.