accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CCH3NCH3CH3

CAS RN 97-84-7

IUPAC Name: [3-(dimethylamino)butyl]dimethylamine
Synonyms: [3-(dimethylamino)butyl]dimethylamine
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: AXFVIWBTKYFOCY-UHFFFAOYNA-N
SMILES: CC(CCN(C)C)N(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results